Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pup-3
Entrez Gene ID
172207
Gene Name
PAP-associated domain-containing protein
Gene Aliases
CELE_F59A3.9
Chromosome Location
Chr.I: 5530616 - 5534649 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059220.5 NP_491621.1 3 - 4 432 76

View Full Product Details