Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lim-7
Entrez Gene ID
172236
Gene Name
LIM domain family
Gene Aliases
CELE_C04F1.3
Chromosome Location
Chr.I: 5715154 - 5725809 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059267.3 NP_491668.1 8 - 9 2316 106
GenBank mRNA U73946.1 - 7 - 8 1013 106

View Full Product Details