The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_059272.6 | NP_491673.1 | 13 - 14 | 1872 | 73 |
GenBank mRNA | AJ277649.1 | - | 13 - 14 | 1780 | 73 |