Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
che-14
Entrez Gene ID
172240
Gene Name
SSD domain-containing protein
Gene Aliases
CELE_F56H1.1
Chromosome Location
Chr.I: 5746633 - 5754306 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059272.6 NP_491673.1 5 - 6 1013 162
GenBank mRNA AJ277649.1 - 5 - 6 921 162

View Full Product Details