Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-2
Entrez Gene ID
172251
Gene Name
Nuclear hormone receptor family member nhr-2
Gene Aliases
CELE_C32F10.6
Chromosome Location
Chr.I: 5815798 - 5821134 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059288.8 NP_491689.4 1 - 2 136 108

View Full Product Details