Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpa-14
Entrez Gene ID
172269
Gene Name
Guanine nucleotide-binding protein alpha-14 subunit
Gene Aliases
CELE_B0207.3
Chromosome Location
Chr.I: 5942390 - 5945651 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025795.3 NP_001020966.2 8 - 9 1197 84

View Full Product Details