Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-31
Entrez Gene ID
177601
Gene Name
protein-coding
Gene Aliases
CELE_C26B2.3
Chromosome Location
Chr.IV: 8022655 - 8029515 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026159.6 NP_001021330.1 7 - 8 1500 77
NM_171391.7 NP_741456.1 6 - 7 1565 77
NR_131580.1 6 - 7 1446 77
GenBank mRNA AF273776.1 - 6 - 7 1555 77

View Full Product Details