Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spp-10
Entrez Gene ID
177651
Gene Name
protein-coding
Gene Aliases
CELE_C28C12.7
Chromosome Location
Chr.IV: 8477176 - 8480656 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171399.6 NP_741465.1 8 - 9 1028 131
NM_171400.5 NP_741466.1 6 - 7 2621 131

View Full Product Details