Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rtel-1
Entrez Gene ID
172946
Gene Name
Regulator of telomere elongation helicase 1 homolog
Gene Aliases
CELE_F25H2.13
Chromosome Location
Chr.I: 10571954 - 10577135 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060368.6 NP_492769.1 11 - 12 2728 153

View Full Product Details