Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-26
Entrez Gene ID
178284
Gene Name
protein-coding
Gene Aliases
CELE_JC8.10
Chromosome Location
Chr.IV: 13261537 - 13272516 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028094.4 NP_001023265.1 3 - 4 547 133
NM_001028095.6 NP_001023266.1 3 - 4 549 133
NM_001129313.3 NP_001122785.1 3 - 4 547 133
NM_001136390.4 NP_001129862.1 3 - 4 544 133
GenBank mRNA AF283322.1 - 3 - 4 569 133
AF283323.1 - 3 - 4 569 133
AF283324.1 - 3 - 4 569 133

View Full Product Details