Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-62
Entrez Gene ID
178845
Gene Name
protein-coding
Gene Aliases
CELE_T28F12.2
Chromosome Location
Chr.V: 4497461 - 4511463 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028998.4 NP_001024169.1 8 - 9 1329 87
NM_001028999.5 NP_001024170.1 8 - 9 1680 87
NM_001029002.4 NP_001024173.1 7 - 8 1202 87
NM_001029004.5 NP_001024175.1 7 - 8 1103 87
NM_001313278.3 NP_001300207.1 6 - 6 1091 87
NM_001380634.1 NP_001367924.1 8 - 9 1317 87
NM_001380635.1 NP_001367923.1 8 - 9 1319 87
NM_001383298.1 7 - 8 2019 87
GenBank mRNA AF427474.1 - 8 - 9 1339 87
AF427475.1 - 8 - 9 1351 87
AF427476.1 - 8 - 9 1522 87
AF427477.1 - 8 - 9 1534 87

View Full Product Details