Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- alh-4
- Entrez Gene ID
-
179026
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_T05H4.13
- Chromosome Location
-
Chr.V: 6427208 - 6429920 on Build WBcel235
- Gene Symbol
- T05H4.11
- Entrez Gene ID
-
179024
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_T05H4.11
- Chromosome Location
-
Chr.V: 6424247 - 6429913 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072231.7 |
NP_504632.1 |
2 - 3 |
643 |
94 |
- Gene Symbol
- atp-4
- Entrez Gene ID
-
179025
- Gene Name
- ATP synthase subunit
- Gene Aliases
-
CELE_T05H4.12
- Chromosome Location
-
Chr.V: 6425860 - 6429916 on Build WBcel235
View Full Product Details