Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ubh-1
- Entrez Gene ID
-
179039
- Gene Name
- Ubiquitin carboxyl-terminal hydrolase
- Gene Aliases
-
CELE_F46E10.8
- Chromosome Location
-
Chr.V: 6508928 - 6511409 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072253.5 |
NP_504654.1 |
7 - 8 |
951 |
67 |
- Gene Symbol
- ubh-2
- Entrez Gene ID
-
24105081
- Gene Name
- Ubiquitin carboxyl-terminal hydrolase
- Gene Aliases
-
CELE_Y40G12A.2
- Chromosome Location
-
Chr.V: 6508907 - 6509817 on Build WBcel235
- Gene Symbol
- ubh-3
- Entrez Gene ID
-
259626
- Gene Name
- Ubiquitin carboxyl-terminal hydrolase
- Gene Aliases
-
CELE_Y40G12A.1
- Chromosome Location
-
Chr.V: 6506754 - 6509817 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072252.7 |
NP_504653.2 |
1 - 2 |
67 |
67 |
View Full Product Details