Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rmd-2
Entrez Gene ID
179391
Gene Name
protein-coding
Gene Aliases
CELE_C27H6.4
Chromosome Location
Chr.V: 9983919 - 9986277 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129394.5 NP_001122866.1 1 - 2 77 100
NM_171522.6 NP_741608.2 5 - 6 536 100
NM_171523.6 NP_741609.1 3 - 4 362 100

View Full Product Details