Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tsp-13
Entrez Gene ID
189737
Gene Name
Tetraspanin
Gene Aliases
CELE_Y39B6A.6
Chromosome Location
Chr.V: 19154106 - 19157718 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171602.3 NP_741692.1 5 - 6 638 107

View Full Product Details