Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hot-2
Entrez Gene ID
180261
Gene Name
protein-coding
Gene Aliases
CELE_Y39B6A.48
Chromosome Location
Chr.V: 19174759 - 19187911 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269917.3 NP_001256846.1 2 - 3 527 110
NM_001269919.3 NP_001256848.1 2 - 3 221 110

View Full Product Details