Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-96
Entrez Gene ID
180413
Gene Name
protein-coding
Gene Aliases
CELE_F13C5.6
Chromosome Location
Chr.X: 611103 - 621290 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171612.7 NP_741703.1 6 - 7 698 83
NM_171939.5 NP_741704.1 6 - 7 698 83

View Full Product Details