Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vps-45
Entrez Gene ID
180449
Gene Name
related to yeast Vacuolar Protein Sorting factor
Gene Aliases
CELE_C44C1.4
Chromosome Location
Chr.X: 970583 - 974848 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171622.6 NP_741714.1 9 - 10 1541 75

View Full Product Details