Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ser-7
Entrez Gene ID
180565
Gene Name
protein-coding
Gene Aliases
CELE_C09B7.1
Chromosome Location
Chr.X: 2673572 - 2682384 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029202.6 NP_001024373.1 3 - 4 916 74
NM_171637.6 NP_741730.1 3 - 4 1045 74
NM_171941.7 NP_741731.1 3 - 4 959 74

View Full Product Details