Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cgef-1
Entrez Gene ID
180571
Gene Name
protein-coding
Gene Aliases
CELE_C14A11.3
Chromosome Location
Chr.X: 2772042 - 2799250 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129633.5 NP_001123105.1 4 - 5 1161 74
NM_171641.6 NP_741736.2 20 - 21 2782 74
NM_171642.6 NP_741737.1 4 - 5 2104 74

View Full Product Details