Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-18
Entrez Gene ID
180671
Gene Name
Homeobox protein ceh-18
Gene Aliases
CELE_ZC64.3
Chromosome Location
Chr.X: 3843361 - 3857797 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171659.6 NP_741758.1 8 - 9 1623 102
GenBank mRNA U16367.1 - 8 - 9 1623 102

View Full Product Details