Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
coel-1
Entrez Gene ID
180700
Gene Name
protein-coding
Gene Aliases
CELE_C52B9.3
Chromosome Location
Chr.X: 4265663 - 4270554 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171665.6 NP_741764.1 1 - 2 175 96
NM_171947.7 NP_741765.1 1 - 2 363 96

View Full Product Details