Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spat-3
Entrez Gene ID
259704
Gene Name
protein-coding
Gene Aliases
CELE_T13H2.5
Chromosome Location
Chr.X: 4596011 - 4605928 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029733.6 NP_001024904.2 3 - 4 565 88
NM_001029734.4 NP_001024905.1 3 - 4 567 88

View Full Product Details