Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbc-7
Entrez Gene ID
180784
Gene Name
protein-coding
Gene Aliases
CELE_C31H2.1
Chromosome Location
Chr.X: 5134280 - 5146744 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373289.3 NP_001359912.1 15 - 16 1647 95
NM_001373290.4 NP_001360675.1 12 - 13 1358 95
NM_171949.8 NP_741779.1 13 - 14 1521 95

View Full Product Details