Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ags-3
Entrez Gene ID
180801
Gene Name
protein-coding
Gene Aliases
CELE_F32A6.4
Chromosome Location
Chr.X: 5289728 - 5303600 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322618.4 NP_001309689.1 1 - 2 84 69
NM_001383577.1 7 - 8 970 69
NM_171684.6 NP_741787.1 7 - 8 894 69
NR_136376.1 7 - 8 954 69
NR_136377.1 5 - 6 800 69

View Full Product Details