Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mec-2
Entrez Gene ID
180826
Gene Name
protein-coding
Gene Aliases
CELE_F14D12.4
Chromosome Location
Chr.X: 5567754 - 5590830 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029395.4 NP_001024566.1 7 - 8 928 87
NM_001029396.4 NP_001024567.1 6 - 7 967 87
NM_001368504.4 NP_001355460.1 2 - 2 682 87
NM_001368505.3 NP_001355457.1 6 - 7 751 87
NM_001368507.2 NP_001355459.1 6 - 7 751 87
NM_001368508.5 NP_001355458.1 8 - 8 990 87
NM_001368511.3 NP_001355478.1 6 - 7 727 87
NM_001368514.4 NP_001355480.1 7 - 7 784 87
NM_001368515.4 NP_001355479.1 6 - 6 703 87
NM_001380971.2 NP_001368710.1 6 - 7 868 87
NM_001380972.1 NP_001368712.1 7 - 8 928 87
NM_001380973.1 NP_001368715.1 7 - 8 928 87
NM_001380974.1 NP_001368713.1 6 - 7 727 87
NM_001380975.1 NP_001368716.1 6 - 7 727 87
NM_001380976.1 NP_001368714.1 6 - 7 703 87
NM_001380977.1 NP_001368717.1 6 - 7 703 87
NM_171691.6 NP_741797.1 7 - 8 971 87
GenBank mRNA U26735.1 - 7 - 8 965 87

View Full Product Details