Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gar-1
Entrez Gene ID
180922
Gene Name
protein-coding
Gene Aliases
CELE_C15B12.5
Chromosome Location
Chr.X: 6479443 - 6484880 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029230.3 NP_001024401.1 8 - 9 819 104
NM_001029232.2 NP_001024403.1 8 - 9 819 104
NM_001373329.1 NP_001360660.1 8 - 9 819 104
GenBank mRNA AF075245.1 - 8 - 9 819 104
AF117301.1 - 8 - 9 819 104

View Full Product Details