Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glr-6
Entrez Gene ID
180955
Gene Name
protein-coding
Gene Aliases
CELE_F41B4.4
Chromosome Location
Chr.X: 6836210 - 6840720 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171712.5 NP_741822.2 3 - 4 467 145

View Full Product Details