Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhx-4
Entrez Gene ID
180964
Gene Name
protein-coding
Gene Aliases
CELE_F14B8.1
Chromosome Location
Chr.X: 6914282 - 6921170 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029391.4 NP_001024562.1 9 - 10 1720 79
NM_001381015.1 NP_001367515.1 9 - 10 1724 79
GenBank mRNA AF497826.1 - 9 - 10 1742 79
AF497827.1 - 6 - 7 1462 79

View Full Product Details