Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-17
Entrez Gene ID
181024
Gene Name
protein-coding
Gene Aliases
CELE_C44E12.3
Chromosome Location
Chr.X: 7599806 - 7608082 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029291.3 NP_001024462.1 2 - 3 221 100
NM_001029292.3 NP_001024463.1 2 - 3 200 100
NM_001029293.3 NP_001024464.1 2 - 3 392 100
NM_001029294.4 NP_001024465.1 2 - 3 415 100
NM_001029295.5 NP_001024466.1 2 - 3 392 100
NM_001029296.8 NP_001024467.1 2 - 3 276 100
NM_001373367.3 NP_001360693.1 1 - 1 200 100
GenBank mRNA AF083643.1 - 2 - 3 200 100
AF083644.1 - 2 - 3 200 100
AF083647.1 - 2 - 3 392 100
AF083648.1 - 2 - 3 392 100
AF083651.1 - 2 - 3 392 100

View Full Product Details