Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-17
Entrez Gene ID
181024
Gene Name
protein-coding
Gene Aliases
CELE_C44E12.3
Chromosome Location
Chr.X: 7599806 - 7608082 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029291.3 NP_001024462.1 7 - 8 1009 136
NM_001029292.3 NP_001024463.1 7 - 8 988 136
NM_001029293.3 NP_001024464.1 7 - 8 1180 136
NM_001029294.4 NP_001024465.1 7 - 8 1203 136
NM_001029295.5 NP_001024466.1 7 - 8 1180 136
NM_001029296.8 NP_001024467.1 7 - 8 1064 136
NM_001373367.3 NP_001360693.1 7 - 7 988 136
GenBank mRNA AF083643.1 - 7 - 8 988 136
AF083644.1 - 7 - 8 988 136
AF083647.1 - 7 - 8 1180 136
AF083648.1 - 7 - 8 1180 136
AF083651.1 - 7 - 8 1180 136

View Full Product Details