Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kin-9
Entrez Gene ID
181098
Gene Name
protein-coding
Gene Aliases
CELE_F08F1.1
Chromosome Location
Chr.X: 8432661 - 8437350 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373393.3 NP_001359981.1 3 - 4 467 103

View Full Product Details