Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lgc-47
Entrez Gene ID
181193
Gene Name
protein-coding
Gene Aliases
CELE_F47A4.1
Chromosome Location
Chr.X: 9801078 - 9807663 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171756.4 NP_741878.1 2 - 3 165 87
NM_171757.7 NP_741879.1 2 - 3 384 87

View Full Product Details