Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-58
Entrez Gene ID
181224
Gene Name
protein-coding
Gene Aliases
CELE_T06H11.1
Chromosome Location
Chr.X: 10104610 - 10133182 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373456.2 NP_001360793.1 11 - 12 1466 93
NM_001373457.2 NP_001360791.1 9 - 10 1346 93
NM_001373458.3 NP_001360790.1 11 - 11 1483 93
NM_001373459.2 NP_001360794.1 8 - 9 1293 93
NM_001373460.3 NP_001360792.1 9 - 10 1203 93
NM_171758.5 NP_741880.1 9 - 10 1327 93
NM_171759.6 NP_741881.1 8 - 9 1391 93

View Full Product Details