Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-48
Entrez Gene ID
181661
Gene Name
protein-coding
Gene Aliases
CELE_ZK662.3
Chromosome Location
Chr.X: 15708213 - 15716875 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001375186.2 NP_001362159.1 3 - 4 326 88
NM_001375187.2 NP_001362160.1 3 - 4 326 88
NM_001375188.2 NP_001361843.1 4 - 5 459 88
NM_001375189.1 NP_001361880.1 4 - 5 459 88

View Full Product Details