Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mec-5
Entrez Gene ID
181678
Gene Name
Uncharacterized protein
Gene Aliases
CELE_E03G2.3
Chromosome Location
Chr.X: 15944009 - 15948955 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171808.5 NP_741941.1 5 - 6 795 67
GenBank mRNA U33919.1 - 5 - 6 817 67

View Full Product Details