Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
egl-44
Entrez Gene ID
174005
Gene Name
protein-coding
Gene Aliases
CELE_F28B12.2
Chromosome Location
Chr.II: 5928802 - 5940830 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356850.3 NP_001343767.1 1 - 1 43 65
NM_171850.7 NP_741007.2 1 - 2 87 65

View Full Product Details