Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clh-1
Entrez Gene ID
174821
Gene Name
protein-coding
Gene Aliases
CELE_T27D12.2
Chromosome Location
Chr.II: 11846507 - 11854745 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027202.7 NP_001022373.1 16 - 17 2606 62
NM_001027203.5 NP_001022374.1 16 - 17 2596 62
GenBank mRNA D84668.1 - 16 - 17 2594 62

View Full Product Details