Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
flap-1
Entrez Gene ID
175964
Gene Name
protein-coding
Gene Aliases
CELE_F57B9.7
Chromosome Location
Chr.III: 6923971 - 6928825 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171879.6 NP_741208.1 2 - 3 203 93

View Full Product Details