Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-32
Entrez Gene ID
176257
Gene Name
protein-coding
Gene Aliases
CELE_ZK637.8
Chromosome Location
Chr.III: 8905615 - 8911756 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027846.2 NP_001023017.1 3 - 4 468 112
NM_001027849.4 NP_001023020.1 3 - 4 468 112
GenBank mRNA AF320899.1 - 3 - 4 466 112
AF320902.1 - 3 - 4 466 112

View Full Product Details