Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-32
Entrez Gene ID
176257
Gene Name
protein-coding
Gene Aliases
CELE_ZK637.8
Chromosome Location
Chr.III: 8905615 - 8911756 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027846.2 NP_001023017.1 5 - 6 840 130
NM_001027847.4 NP_001023018.1 5 - 6 792 130
NM_001027848.6 NP_001023019.1 5 - 6 807 130
NM_001027849.4 NP_001023020.1 5 - 6 840 130
NM_001027850.5 NP_001023021.1 5 - 6 807 130
NM_001027851.2 NP_001023022.1 5 - 6 792 130
GenBank mRNA AF320899.1 - 5 - 6 838 130
AF320900.1 - 5 - 6 790 130
AF320901.1 - 5 - 6 805 130
AF320902.1 - 5 - 6 838 130
AF320903.1 - 5 - 6 805 130
AF320904.1 - 5 - 6 790 130

View Full Product Details