Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-32
Entrez Gene ID
176257
Gene Name
protein-coding
Gene Aliases
CELE_ZK637.8
Chromosome Location
Chr.III: 8905615 - 8911756 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027846.2 NP_001023017.1 7 - 8 2233 95
NM_001027848.6 NP_001023019.1 7 - 8 2200 95
NM_001027851.2 NP_001023022.1 7 - 8 2185 95
GenBank mRNA AF320899.1 - 7 - 8 2231 95
AF320901.1 - 7 - 8 2198 95
AF320904.1 - 7 - 8 2183 95

View Full Product Details