Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-32
Entrez Gene ID
176257
Gene Name
protein-coding
Gene Aliases
CELE_ZK637.8
Chromosome Location
Chr.III: 8905615 - 8911756 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027847.4 NP_001023018.1 3 - 4 474 117
NM_001027851.2 NP_001023022.1 3 - 4 474 117
GenBank mRNA AF320900.1 - 3 - 4 472 117
AF320904.1 - 3 - 4 472 117

View Full Product Details