Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-32
Entrez Gene ID
176257
Gene Name
protein-coding
Gene Aliases
CELE_ZK637.8
Chromosome Location
Chr.III: 8905615 - 8911756 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027847.4 NP_001023018.1 7 - 8 2169 96
NM_001027849.4 NP_001023020.1 7 - 8 2217 96
NM_001027850.5 NP_001023021.1 7 - 8 2184 96
GenBank mRNA AF320900.1 - 7 - 8 2167 96
AF320902.1 - 7 - 8 2215 96
AF320903.1 - 7 - 8 2182 96

View Full Product Details