Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pxf-1
Entrez Gene ID
177894
Gene Name
protein-coding
Gene Aliases
CELE_T14G10.2
Chromosome Location
Chr.IV: 10164288 - 10174341 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028218.6 NP_001023389.1 1 - 2 98 67
NM_001028219.3 NP_001023390.1 1 - 2 98 67
GenBank mRNA AF170796.1 - 1 - 2 87 67
AF308447.1 - 1 - 2 87 67
AF308448.1 - 1 - 2 87 67

View Full Product Details