Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-96
Entrez Gene ID
180413
Gene Name
protein-coding
Gene Aliases
CELE_F13C5.6
Chromosome Location
Chr.X: 611103 - 621290 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171939.5 NP_741704.1 11 - 12 1187 64

View Full Product Details