Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-84
Entrez Gene ID
181480
Gene Name
protein-coding
Gene Aliases
CELE_F54B11.3
Chromosome Location
Chr.X: 13584775 - 13589827 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029536.5 NP_001024707.1 6 - 7 2165 129
NM_001381134.1 NP_001366990.1 6 - 7 2165 129
GenBank mRNA AF200706.1 - 6 - 7 2172 129
AF200707.1 - 6 - 7 2170 129

View Full Product Details