Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ser-2
Entrez Gene ID
182110
Gene Name
protein-coding
Gene Aliases
CELE_C02D4.2
Chromosome Location
Chr.X: 15286533 - 15302963 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029168.4 NP_001024339.1 8 - 9 935 72

View Full Product Details