Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gpdh-2
Entrez Gene ID
176399
Gene Name
protein-coding
Gene Aliases
CELE_K11H3.1
Chromosome Location
Chr.III: 9853807 - 9859183 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181903.7 NP_871632.1 1 - 2 119 87

View Full Product Details