Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tlk-1
Entrez Gene ID
176369
Gene Name
protein-coding
Gene Aliases
CELE_C07A9.3
Chromosome Location
Chr.III: 9699534 - 9710315 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268088.3 NP_001255017.1 6 - 7 2582 85
NM_181913.5 NP_871642.2 8 - 9 2825 85

View Full Product Details